Assay Details
| Gene Symbol: Camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286809.1 | NP_001273738.1 | 4 - 5 | 635 | 61 | |
| NM_009792.3 | NP_033922.1 | 5 - 6 | 668 | 61 | ||
| NM_177407.4 | NP_803126.1 | 16 - 17 | 1356 | 61 | ||
| GenBank mRNA | AK083245.1 | - | 16 - 17 | 1357 | 61 | |
| AK122410.2 | - | 16 - 17 | 1352 | 61 | ||
| BB659027.1 | - | 4 - 5 | 591 | 61 | ||
| BC031745.1 | - | 16 - 17 | 1337 | 4506005 | 61 | |
More Information
Set Membership: |
|

Camk2a
Genomic Map