Assay Details
| Gene Symbol: Cdh22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174988.3 | NP_778153.2 | 3 - 4 | 1036 | 54 | |
| GenBank mRNA | AK140248.1 | - | 3 - 4 | 1154 | 54 | |
| BC112407.1 | - | 2 - 3 | 731 | 40060660 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh22
Genomic Map