Assay Details
| Gene Symbol: Sox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011437.1 | NP_035567.1 | 3 - 4 | 986 | 114 | |
| GenBank mRNA | AF017182.1 | - | 4 - 5 | 1048 | 114 | |
| AF047043.1 | - | 2 - 3 | 752 | 114 | ||
| AF047389.1 | - | 2 - 3 | 753 | 114 | ||
| AK042542.1 | - | 3 - 4 | 988 | 114 | ||
| AK043220.1 | - | 3 - 4 | 983 | 114 | ||
| BC018551.1 | - | 2 - 3 | 784 | 3675437 | 114 | |
| BC023356.1 | - | 3 - 4 | 820 | 5027211 | 114 | |
| BC025171.1 | - | 3 - 4 | 949 | 4912940 | 114 | |
| BC051058.1 | - | 3 - 4 | 820 | 114 | ||
| U66141.1 | - | 3 - 4 | 983 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
neural crest cell migration
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
peripheral nervous system development
positive regulation of gliogenesis
cell differentiation
melanocyte differentiation
lacrimal gland development
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
enteric nervous system development
digestive tract morphogenesis
developmental growth
oligodendrocyte differentiation
morphogenesis of a branching epithelium
negative regulation of canonical Wnt signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
identical protein binding
transcription regulatory region DNA binding
in utero embryonic development
neural crest cell migration
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
peripheral nervous system development
positive regulation of gliogenesis
cell differentiation
melanocyte differentiation
lacrimal gland development
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
enteric nervous system development
digestive tract morphogenesis
developmental growth
oligodendrocyte differentiation
morphogenesis of a branching epithelium
negative regulation of canonical Wnt signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
identical protein binding
transcription regulatory region DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364763 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364764 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364762 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364761 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094729 | U66141.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094729 | BC025171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094729 | BC051058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094729 | D87031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094729 | BC018551.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932411 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916685 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 791 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107072 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136803 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094729 | U66141.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094729 | BC025171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094729 | BC051058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094729 | D87031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094729 | BC018551.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P346704 | U66141.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P346704 | BC051058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P346704 | D87031.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P346704 | BC023356.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P346704 | AK042542.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192625 | U66141.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192625 | BC025171.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192625 | BC051058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192625 | BC018551.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192625 | BC023356.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08429563 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107072 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136803 | - | - |
| Whole Mouse Genome G4122A | A_51_P346704 | U66141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346704 | BC051058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346704 | D87031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346704 | BC023356.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P346704 | AK042542.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192625 | U66141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192625 | BC025171.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192625 | BC051058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192625 | BC018551.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192625 | BC023356.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107072 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136803 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364763 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364761 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364762 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30364764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12077580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11952971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12079756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12079751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11970944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12435749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12435748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12435758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12032187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12146617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11952973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12146620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12079748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11952981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12077583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12077582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30364800 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430462 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430460 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430465 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424985_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451689_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425369_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430458 | - | - |
| Mouse Genome 430 2.0 Array | 1424985_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425369_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451689_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451689_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424985_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425369_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20665_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20665_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014536.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014536.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014536.mm.1 | TC1500001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014539.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014539.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014539.mm.1 | TC1500001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014543.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014543.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014543.mm.1 | TC1500001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014538.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014538.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014538.mm.1 | TC1500001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014535.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014535.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014535.mm.1 | TC1500001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014537.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014537.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014537.mm.1 | TC1500001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014542.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014542.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014542.mm.1 | TC1500001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500014541.mm.1 | NM_011437.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014541.mm.1 | NM_011437.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014541.mm.1 | TC1500001863.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424985_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425369_a_at | - | - |
| Mouse Expression Set 430 | 1425369_PM_a_at | - | - |
| Mouse Expression Set 430 | 1424985_a_at | - | - |
| Mouse Expression Set 430 | 1451689_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451689_a_at | - | - |
| Murine Genome U74v2 Set | 100283_PM_at | - | - |
| Murine Genome U74v2 Set | 102856_PM_at | - | - |
| Murine Genome U74v2 Set | 102856_at | - | - |
| Murine Genome U74v2 Set | 100283_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425369_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425369_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424985_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424985_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451689_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451689_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424985_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425369_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424985_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451689_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451689_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425369_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228105 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228105 | - | - |

Sox10
Genomic Map