Assay Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178060.3 | NP_835161.1 | 3 - 4 | 556 | 63 | |
| GenBank mRNA | AK078233.1 | - | 3 - 4 | 556 | 63 | |
| BC046795.1 | - | 528 | 5717946 | 63 | ||
| X07750.1 | - | 2 - 3 | 126 | 63 | ||
| X07751.1 | - | 126 | 63 | |||
| X07752.1 | - | 126 | 63 | |||
| X51983.1 | - | 2 - 3 | 130 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thra
Genomic Map