Assay Details
| Gene Symbol: Igf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010514.3 | NP_034644.2 | 1 - 2 | 436 | 88 | |
| GenBank mRNA | CF742200.1 | - | 1 - 2 | 295 | 88 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_010514.3 transcripts.
Set Membership: |
|

Igf2
Genomic Map