Assay Details
Gene Symbol: Lim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177693.3 | NP_808361.1 | 4 - 5 | 523 | 55 | |
GenBank mRNA | AK029354.1 | - | 4 - 5 | 524 | 55 | |
AK164958.1 | - | 4 - 5 | 522 | 55 | ||
BC092386.1 | - | 4 - 5 | 536 | 30551056 | 55 | |
BC119253.1 | - | 4 - 5 | 504 | 8734002 | 55 | |
BC119255.1 | - | 4 - 5 | 504 | 8734004 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | BC092386.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | AK164958.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | AK029354.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | BC092386.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | AK164958.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487028 | AK029354.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537108 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917215 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121700 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174614 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | BC092386.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | AK164958.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | AK029354.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | BC092386.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | AK164958.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487028 | AK029354.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P135653 | AK145594.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | BC092386.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | AK164958.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | AK029354.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | BC092386.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | AK164958.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P487028 | AK029354.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082293 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082312 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082311 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082305 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082301 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082317 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082306 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082289 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082298 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082283 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082308 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082320 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082321 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082319 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082310 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082314 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082309 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082300 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082292 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082296 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25082278 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174614 | - | - |
Whole Mouse Genome G4122A | A_51_P487028 | BC092386.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P487028 | AK164958.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P487028 | AK029354.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P487028 | BC092386.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P487028 | AK164958.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P487028 | NM_177693.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P487028 | AK029354.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113032 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127937 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174614 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06475535 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06475540 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06475533 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06475542 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06475541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082304 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06056936 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082311 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06475534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06634448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05996971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082314 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06573126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082312 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06591781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06594845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082289 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06659565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06475539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05996969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06475532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082254 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10552403 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552402 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552404 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552400 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552405 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552399 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438323_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10552398 | - | - |
Mouse Genome 430 2.0 Array | 1438323_at | - | - |
Mouse Genome 430 2.0 Array | 1438323_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233187_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233187_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700006138.mm.1 | TC0700000758.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006138.mm.1 | NM_177693.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006138.mm.1 | NM_177693.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006137.mm.1 | TC0700000758.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006137.mm.1 | NM_177693.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006137.mm.1 | NM_177693.3 | Refseq |
Mouse Expression Set 430 | 1438323_PM_at | - | - |
Mouse Expression Set 430 | 1438323_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438323_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438323_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6980162 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006980162 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247117 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1247117 | - | - |
mouse-6 v1.1 (Illumina) | scl32757.5.1_2-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6980162 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32757.5.1_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980162 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1247117 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247117 | - | - |
