| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700002550.mm.1 |
NM_027937.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002550.mm.1 |
NM_027937.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002550.mm.1 |
TC1700000341.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002549.mm.1 |
NM_027937.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002549.mm.1 |
NM_027937.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002549.mm.1 |
TC1700000341.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002548.mm.1 |
NM_027937.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002548.mm.1 |
NM_027937.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002548.mm.1 |
TC1700000341.mm.1 |
MTA TC |