Assay Details
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010157.3 | NP_034287.3 | 7 - 8 | 1630 | 83 | |
NM_207707.1 | NP_997590.1 | 8 - 9 | 1684 | 83 | ||
NR_104386.1 | 6 - 7 | 1491 | 83 | |||
GenBank mRNA | AK054290.1 | - | 8 - 9 | 1682 | 83 | |
AK054301.1 | - | 6 - 7 | 1492 | 83 | ||
AK054413.1 | - | 8 - 9 | 1685 | 83 | ||
AY054413.1 | - | 7 - 8 | 1145 | 83 | ||
BC141075.1 | - | 6 - 7 | 1332 | 9055687 | 83 | |
BC145329.1 | - | 6 - 7 | 1332 | 9053885 | 83 | |
U81451.1 | - | 6 - 7 | 1102 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
