Assay Details
Gene Symbol: Prl7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011120.2 | NP_035250.1 | 5 - 6 | 606 | 103 | |
GenBank mRNA | AK005421.1 | - | 5 - 6 | 603 | 103 | |
AK005557.1 | - | 5 - 6 | 607 | 104 | ||
AK019461.1 | - | 5 - 6 | 606 | 103 | ||
BC099470.1 | - | 5 - 6 | 595 | 30787403 | 103 | |
X02594.1 | - | 5 - 6 | 606 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | NM_011120.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | AK005421.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | AK005557.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | AK019461.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | BC099470.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | NM_011120.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | AK005421.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | AK005557.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | AK019461.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P292196 | BC099470.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46367 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973518 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725974 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26477 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26820 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125748 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | NM_011120.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | AK005421.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | AK005557.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | AK019461.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | BC099470.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | NM_011120.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | AK005421.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | AK005557.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | AK019461.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P292196 | BC099470.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903339 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903345 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903358 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903365 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903336 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903370 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903367 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903354 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903368 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903361 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903352 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07819132 | - | - |
Whole Mouse Genome G4122A | A_51_P292196 | NM_011120.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | AK005421.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | AK005557.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | AK019461.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | BC099470.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | NM_011120.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | AK005421.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | AK005557.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | AK019461.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P292196 | BC099470.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116925 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100157 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10863462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903356 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903367 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903358 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903365 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10557585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903344 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903310 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903371 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903363 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903370 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903350 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903342 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903346 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903336 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903354 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903322 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408440 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408438 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408442 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408441 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408443 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408439 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1460217_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408436 | - | - |
Mouse Genome 430 2.0 Array | 1460217_at | - | - |
Mouse Genome 430 2.0 Array | 1460217_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18814_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18814_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300010571.mm.1 | NM_011120.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010571.mm.1 | NM_011120.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010571.mm.1 | TC1300001698.mm.1 | MTA TC |
Mouse Expression Set 430 | 1460217_PM_at | - | - |
Mouse Expression Set 430 | 1460217_at | - | - |
Murine Genome U74v2 Set | 98023_r_at | - | - |
Murine Genome U74v2 Set | 98023_PM_r_at | - | - |
Murine Genome U74v2 Set | 171445_PM_i_at | - | - |
Murine Genome U74v2 Set | 169924_r_at | - | - |
Murine Genome U74v2 Set | 171445_i_at | - | - |
Murine Genome U74v2 Set | 169924_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1460217_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1460217_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460217_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460217_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 678863 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3990204 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3990204 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003990204 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215580 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1215580 | - | - |
mouse-6 v1.1 (Illumina) | scl018814.2_21-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3990204 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl018814.2_21-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990204 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1215580 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215580 | - | - |
