Assay Details
| Gene Symbol: Prrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011127.2 | NP_035257.1 | 3 - 4 | 1613 | 107 | |
| GenBank mRNA | AB221686.1 | - | 3 - 4 | 598 | 107 | |
| X59725.1 | - | 3 - 4 | 739 | 107 | ||
More Information
Set Membership: |
|

Prrx1
Genomic Map