Assay Details
Gene Symbol: Prl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011164.2 | NP_035294.2 | 1 - 2 | 91 | 88 | |
GenBank mRNA | AK017521.1 | - | 1 - 2 | 85 | 88 | |
AK017579.1 | - | 1 - 2 | 85 | 88 | ||
AK077493.1 | - | 1 - 2 | 88 | 88 | ||
AK133736.1 | - | 1 - 2 | 85 | 88 | ||
AK133752.1 | - | 1 - 2 | 85 | 88 | ||
AK161557.1 | - | 1 - 2 | 85 | 88 | ||
AK161565.1 | - | 1 - 2 | 85 | 88 | ||
AK161619.1 | - | 1 - 2 | 89 | 88 | ||
BC061141.1 | - | 1 - 2 | 84 | 30250937 | 88 | |
X02892.1 | - | 1 - 2 | 83 | 88 | ||
X04418.1 | - | 1 - 2 | 85 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK017521.1, AK017579.1, AK077493.1, AK133736.1, AK133752.1, AK161557.1, AK161565.1, AK161619.1, BC061141.1, NM_011164.2, X02892.1, X04418.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
