Assay Details
| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011240.3 | NP_035370.2 | 2 - 3 | 259 | 120 | |
| GenBank mRNA | AK041932.1 | - | 2 - 3 | 192 | 120 | |
| AK145306.1 | - | 2 - 3 | 190 | 120 | ||
More Information
Set Membership: |
|

Ranbp2
Genomic Map