Assay Details
| Gene Symbol: Dpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291078.1 | NP_001278007.1 | 5 - 6 | 656 | 72 | |
| NM_011262.5 | NP_035392.1 | 5 - 6 | 656 | 72 | ||
| GenBank mRNA | AK004812.1 | - | 5 - 6 | 593 | 72 | |
| AK138047.1 | - | 5 - 6 | 590 | 72 | ||
| AK144106.1 | - | 5 - 6 | 595 | 72 | ||
| AK144848.1 | - | 5 - 6 | 591 | 72 | ||
| AK144954.1 | - | 5 - 6 | 602 | 72 | ||
| AK145851.1 | - | 5 - 6 | 652 | 72 | ||
| AK156252.1 | - | 4 - 5 | 426 | 72 | ||
| AK186034.1 | - | 1 - 2 | 92 | 72 | ||
| BC012709.1 | - | 5 - 6 | 586 | 4208205 | 72 | |
| CX207082.1 | - | 5 - 6 | 73 | |||
| U43921.1 | - | 5 - 6 | 608 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpf2
Genomic Map