Assay Details
| Gene Symbol: Aoah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012054.4 | NP_036184.1 | 11 - 12 | 1120 | 58 | |
| GenBank mRNA | AK084452.1 | - | 11 - 12 | 1115 | 58 | |
| AK085343.1 | - | 11 - 12 | 1092 | 58 | ||
| AK133301.1 | - | 11 - 12 | 1061 | 58 | ||
| AK170293.1 | - | 10 - 11 | 656 | 58 | ||
| AK171572.1 | - | 11 - 12 | 939 | 58 | ||
| BC150767.1 | - | 11 - 12 | 914 | 9087678 | 58 | |
| CT010242.1 | - | 11 - 12 | 839 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aoah
Genomic Map