Assay Details
| Gene Symbol: Aoah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012054.4 | NP_036184.1 | 17 - 18 | 1647 | 68 | |
| GenBank mRNA | AK084452.1 | - | 17 - 18 | 1642 | 68 | |
| AK085343.1 | - | 17 - 18 | 1619 | 68 | ||
| AK133301.1 | - | 17 - 18 | 1588 | 68 | ||
| AK170293.1 | - | 16 - 17 | 1183 | 68 | ||
| AK171572.1 | - | 17 - 18 | 1466 | 68 | ||
| BC150767.1 | - | 17 - 18 | 1441 | 9087678 | 68 | |
| CT010242.1 | - | 17 - 18 | 1366 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aoah
Genomic Map