Assay Details
| Gene Symbol: Gfpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013528.3 | NP_038556.1 | 9 - 10 | 942 | 86 | |
| GenBank mRNA | AF334736.1 | - | 10 - 11 | 947 | 86 | |
| AK019852.1 | - | 9 - 10 | 938 | 86 | ||
| BC002283.1 | - | 6 - 7 | 541 | 86 | ||
| BC010516.1 | - | 9 - 10 | 890 | 4191553 | 86 | |
| BC050762.1 | - | 9 - 10 | 938 | 6742987 | 86 | |
| U00932.1 | - | 8 - 9 | 789 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gfpt1
Genomic Map