Assay Details
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013739.2 | NP_038767.1 | 3 - 4 | 496 | 130 | |
GenBank mRNA | AF179242.1 | - | 3 - 4 | 496 | 130 | |
AF237580.1 | - | 3 - 4 | 492 | 130 | ||
AK137520.1 | - | 3 - 4 | 483 | 130 | ||
AK154412.1 | - | 2 - 3 | 1319 | 130 | ||
AK154750.1 | - | 2 - 3 | 1319 | 130 | ||
AK155180.1 | - | 2 - 3 | 1326 | 130 | ||
AK156060.1 | - | 2 - 3 | 1326 | 130 | ||
AK172511.1 | - | 2 - 3 | 1326 | 130 | ||
BC048784.1 | - | 2 - 3 | 1627 | 130 | ||
BC116922.1 | - | 3 - 4 | 495 | 40126241 | 130 | |
BC120535.1 | - | 3 - 4 | 495 | 40129458 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
