Assay Details
| Gene Symbol: Il21r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021887.2 | NP_068687.1 | 4 - 5 | 792 | 70 | |
| GenBank mRNA | AF254068.1 | - | 4 - 5 | 496 | 70 | |
| AF269134.1 | - | 3 - 4 | 354 | 70 | ||
| AF279436.1 | - | 4 - 5 | 744 | 70 | ||
| AF477982.1 | - | 3 - 4 | 370 | 70 | ||
| AF477983.1 | - | 3 - 4 | 370 | 70 | ||
| AF477984.1 | - | 3 - 4 | 370 | 70 | ||
| AF477985.1 | - | 3 - 4 | 370 | 70 | ||
| AK040073.1 | - | 3 - 4 | 690 | 70 | ||
| AK150824.1 | - | 4 - 5 | 753 | 70 | ||
| AK171826.1 | - | 4 - 5 | 731 | 70 | ||
| AK172032.1 | - | 4 - 5 | 764 | 70 | ||
| BC057861.1 | - | 4 - 5 | 741 | 5315350 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il21r
Genomic Map