Assay Details
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148938.3 | NP_683740.1 | 2 - 3 | 751 | 55 | |
GenBank mRNA | AF330257.1 | - | 1 - 2 | 251 | 55 | |
AK031831.1 | - | 2 - 3 | 411 | 55 | ||
AK040798.1 | - | 2 - 3 | 411 | 55 | ||
AK045716.1 | - | 2 - 3 | 361 | 55 | ||
AK045949.1 | - | 2 - 3 | 430 | 55 | ||
AK049090.1 | - | 2 - 3 | 411 | 55 | ||
AK080879.1 | - | 2 - 3 | 434 | 55 | ||
AK082736.1 | - | 2 - 3 | 411 | 55 | ||
AK084968.1 | - | 2 - 3 | 395 | 55 | ||
AK158777.1 | - | 1 - 2 | 285 | 55 | ||
BC028638.1 | - | 2 - 3 | 353 | 55 | ||
BC058711.1 | - | 2 - 3 | 674 | 6842500 | 55 | |
BC066154.1 | - | 2 - 3 | 354 | 30102325 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
