Assay Details
| Gene Symbol: Cherp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138585.3 | NP_613051.3 | 11 - 12 | 2158 | 80 | |
| GenBank mRNA | AF536541.1 | - | 11 - 12 | 2060 | 80 | |
| AK049765.1 | - | 11 - 12 | 2069 | 80 | ||
| AK144328.1 | - | 11 - 12 | 2081 | 80 | ||
| AK171907.1 | - | 4 - 5 | 1134 | 80 | ||
| AK210033.1 | - | 1 - 2 | 102 | 80 | ||
| BC032191.1 | - | 11 - 12 | 2076 | 5322208 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cherp
Genomic Map