Assay Details
| Gene Symbol: Rab2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172601.3 | NP_766189.1 | 3 - 4 | 340 | 84 | |
| GenBank mRNA | AB232586.1 | - | 3 - 4 | 186 | 84 | |
| AK005230.1 | - | 3 - 4 | 243 | 84 | ||
| AK038393.1 | - | 3 - 4 | 275 | 84 | ||
| AK044202.1 | - | 3 - 4 | 217 | 84 | ||
| AK160249.1 | - | 3 - 4 | 491 | 84 | ||
| AK160351.1 | - | 3 - 4 | 206 | 84 | ||
| AK169657.1 | - | 3 - 4 | 284 | 84 | ||
| BC033312.1 | - | 3 - 4 | 237 | 4481516 | 84 | |
| BC046334.1 | - | 3 - 4 | 282 | 6492838 | 84 | |
| BC054719.1 | - | 3 - 4 | 273 | 84 | ||
| BY020048.1 | - | 3 - 4 | 341 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab2b
Genomic Map