Assay Details
| Gene Symbol: AI504432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033498.1 | 1 - 2 | 70 | 63 | ||
| GenBank mRNA | AK039146.1 | - | 1 - 2 | 71 | 63 | |
| BC127040.1 | - | 1 - 2 | 71 | 63 | ||
| BC127041.1 | - | 1 - 2 | 71 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018176 | AK039146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018176 | AK046860.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018176 | NR_033498.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109564 | AK039146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109564 | AK046860.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109564 | NR_033498.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176989 | - | - |
| Whole Mouse Genome G4122A | A_52_P683366 | AK039146.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P683366 | AK046860.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P683366 | NR_033498.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P679247 | AK030783.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P679247 | AK040017.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P679247 | AK046860.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P679247 | NR_033498.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03060203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02704092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22403841 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10495187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495188 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456064_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455161_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495186 | - | - |
| Mouse Genome 430 2.0 Array | 1456064_at | - | - |
| Mouse Genome 430 2.0 Array | 1456064_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455161_at | - | - |
| Mouse Genome 430 2.0 Array | 1455161_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300009042.mm.1 | NR_033498.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009042.mm.1 | NR_033498.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009042.mm.1 | TC0300001110.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009041.mm.1 | NR_033498.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009041.mm.1 | NR_033498.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009041.mm.1 | TC0300001110.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009043.mm.1 | NR_033498.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009043.mm.1 | NR_033498.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009043.mm.1 | TC0300001110.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456064_at | - | - |
| Mouse Expression Set 430 | 1456064_PM_at | - | - |
| Mouse Expression Set 430 | 1455161_PM_at | - | - |
| Mouse Expression Set 430 | 1455161_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456064_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456064_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456064_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456064_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455161_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455161_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 401243 | - | - |
| ABI Mouse Genome Survey Microarray | 765576 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100050020 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2606331 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2606331 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2606331 | - | - |

AI504432
Genomic Map