Assay Details
| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173788.3 | NP_776149.1 | 15 - 16 | 2622 | 69 | |
| GenBank mRNA | AK158195.1 | - | 10 - 11 | 1173 | 69 | |
| BC023420.1 | - | 2 - 3 | 263 | 69 | ||
| BC037458.1 | - | 10 - 11 | 1078 | 69 | ||
| BC042470.1 | - | 15 - 16 | 2622 | 5137261 | 69 | |
| BC049896.1 | - | 5 - 6 | 717 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr2
Genomic Map