Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500002455.mm.1 |
TC0500000303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002455.mm.1 |
NM_001048192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002455.mm.1 |
NM_001048192.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002455.mm.1 |
NM_174849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002455.mm.1 |
NM_174849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002454.mm.1 |
TC0500000303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002454.mm.1 |
NM_001048192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002454.mm.1 |
NM_001048192.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002454.mm.1 |
NM_174849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002454.mm.1 |
NM_174849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002457.mm.1 |
TC0500000303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002457.mm.1 |
NM_001048192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002457.mm.1 |
NM_001048192.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002457.mm.1 |
NM_174849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002457.mm.1 |
NM_174849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002442.mm.1 |
TC0500000303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002442.mm.1 |
NM_174849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002442.mm.1 |
NM_174849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002449.mm.1 |
TC0500000303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002449.mm.1 |
NM_001048192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002449.mm.1 |
NM_001048192.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002458.mm.1 |
TC0500000303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500002458.mm.1 |
NM_001048192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500002458.mm.1 |
NM_001048192.2 |
Refseq |