Assay Details
Gene Symbol: Agbl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048192.2 | NP_001041657.1 | 7 - 8 | 1117 | 120 | |
NM_001294262.1 | NP_001281191.1 | 6 - 7 | 1145 | 120 | ||
NM_174849.3 | NP_777274.1 | 6 - 7 | 1034 | 120 | ||
NR_122116.1 | 6 - 7 | 1101 | 120 | |||
NR_122117.1 | 6 - 7 | 1101 | 120 | |||
GenBank mRNA | AK029676.1 | - | 6 - 7 | 1035 | 120 | |
AK035680.1 | - | 4 - 5 | 602 | 120 | ||
AK044195.1 | - | 5 - 6 | 2439 | 120 | ||
AK083656.1 | - | 6 - 7 | 984 | 120 | ||
BC057349.1 | - | 6 - 7 | 1035 | 6813901 | 120 | |
DQ867034.1 | - | 6 - 7 | 1030 | 120 | ||
DQ867035.1 | - | 6 - 7 | 1030 | 120 | ||
DQ867036.1 | - | 7 - 8 | 1117 | 120 | ||
DQ867037.1 | - | 6 - 7 | 1030 | 120 | ||
DQ867038.1 | - | 6 - 7 | 1030 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
