Assay Details
| Gene Symbol: Rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175109.3 | NP_780318.1 | 3 - 4 | 312 | 55 | |
| GenBank mRNA | AB233218.1 | - | 3 - 4 | 308 | 55 | |
| AK010954.1 | - | 3 - 4 | 296 | 55 | ||
| AK075974.1 | - | 3 - 4 | 293 | 55 | ||
| BC038691.1 | - | 3 - 4 | 298 | 1495043 | 55 | |
| BC050115.1 | - | 3 - 4 | 301 | 30017673 | 55 | |
| BC065381.1 | - | 2 - 3 | 245 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371153 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371159 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371158 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | NM_175109.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | AB233218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | AK075974.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | BC038691.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | BC050115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | NM_175109.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | AB233218.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | AK075974.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | BC038691.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P176505 | BC050115.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147900 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | NM_175109.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | AB233218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | AK075974.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | BC038691.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | BC050115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | NM_175109.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | AB233218.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | AK075974.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | BC038691.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P176505 | BC050115.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30371174 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08432443 | - | - |
| Whole Mouse Genome G4122A | A_52_P474797 | NM_175109.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474797 | AB233218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474797 | AK010954.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474797 | AK075974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P474797 | BC038691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | NM_175109.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | AB233218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | AK075974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | BC038691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | BC050115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | NM_175109.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | AB233218.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | AK075974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | BC038691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176505 | BC050115.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122701 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12350693 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12350696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12177927 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371159 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30371153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30371201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12241707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12305424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12455723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11912786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12238930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12172347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12212858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12350695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12238931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12076530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12172344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11883658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12462575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30371191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11912787 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430685 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430682 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430683 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430684 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455174_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430681 | - | - |
| Mouse Genome 430 2.0 Array | 1455174_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455174_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66538_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66538_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500014967.mm.1 | NM_175109.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014967.mm.1 | NM_175109.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500014967.mm.1 | TC1500001905.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455174_at | - | - |
| Mouse Expression Set 430 | 1455174_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455174_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455174_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 912671 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2480075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2480075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480075 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2699989 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2699989 | - | - |
| mouse-6 v1.1 (Illumina) | scl46945.2.7_65-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46945.2.7_65-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480075 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2699989 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2699989 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl46945.2.7_65-S | - | - |

Rps19bp1
Genomic Map