Assay Details
| Gene Symbol: 2810410L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK081626.1 | - | 2125 | 83 | ||
| Gene Symbol: Gm9734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038227.1 | - | 1 | 83 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | AK162474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | AK164256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | NR_030682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | AK013071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | AK081626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | AK162474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | NR_030682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | AK013071.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882515 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | AK081626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | AK162474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | NR_030682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | AK013071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | AK162474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | AK164256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | NR_030682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | AK013071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P626035 | AK081626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P626035 | AK038227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59285 | AK162474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59285 | AK013071.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09185603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09185606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09198963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09328509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09580839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09124925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182970 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1438972_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429510_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393749 | - | - |
| Mouse Genome 430 2.0 Array | 1438972_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429510_at | - | - |
| Mouse Genome 430 2.0 Array | 1429510_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438972_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100042332_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100042332_at | - | - |
| Mouse Expression Set 430 | 1429510_PM_at | - | - |
| Mouse Expression Set 430 | 1438972_x_at | - | - |
| Mouse Expression Set 430 | 1438972_PM_x_at | - | - |
| Mouse Expression Set 430 | 1429510_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438972_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429510_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438972_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429510_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 638132 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 520722 | - | - |

2810410L24Rik
Genomic Map