Assay Details
| Gene Symbol: Bora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175265.4 | NP_780474.1 | 7 - 8 | 826 | 96 | |
| GenBank mRNA | AK034930.1 | - | 7 - 8 | 827 | 96 | |
| AK141389.1 | - | 7 - 8 | 777 | 96 | ||
| AK167430.1 | - | 7 - 8 | 758 | 96 | ||
| BC052183.1 | - | 7 - 8 | 727 | 6336038 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bora
Genomic Map