| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200013863.mm.1 |
NM_175285.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013863.mm.1 |
NM_175285.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013863.mm.1 |
TC0200001801.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200013862.mm.1 |
NM_175285.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013862.mm.1 |
NM_175285.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013862.mm.1 |
TC0200001801.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200013864.mm.1 |
NM_175285.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013864.mm.1 |
NM_175285.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013864.mm.1 |
TC0200001801.mm.1 |
MTA TC |