Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175300.4 | NP_780509.2 | 2 - 3 | 835 | 100 | |
| GenBank mRNA | AK033784.1 | - | 2 - 3 | 803 | 100 | |
| AK129351.1 | - | 2 - 3 | 788 | 100 | ||
| AK154465.1 | - | 2 - 3 | 789 | 100 | ||
| AK170676.1 | - | 2 - 3 | 801 | 100 | ||
| BC027351.1 | - | 2 - 3 | 783 | 100 | ||
| BC038267.1 | - | 1 - 2 | 147 | 100 | ||
| BC096381.1 | - | 1 - 2 | 1028 | 100 | ||
| BC137583.1 | - | 2 - 3 | 803 | 8860602 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map