Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175300.4 | NP_780509.2 | 6 - 7 | 1382 | 85 | |
| GenBank mRNA | AK033784.1 | - | 6 - 7 | 1350 | 85 | |
| AK129351.1 | - | 6 - 7 | 1335 | 85 | ||
| AK154465.1 | - | 6 - 7 | 1336 | 85 | ||
| AK170676.1 | - | 6 - 7 | 1348 | 85 | ||
| AK204259.1 | - | 1 - 2 | 66 | 85 | ||
| BC027351.1 | - | 6 - 7 | 1330 | 85 | ||
| BC038267.1 | - | 5 - 6 | 694 | 85 | ||
| BC096381.1 | - | 5 - 6 | 1575 | 85 | ||
| BC137583.1 | - | 6 - 7 | 1350 | 8860602 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map