Assay Details
| Gene Symbol: Eogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175313.4 | NP_780522.1 | 13 - 14 | 1585 | 66 | |
| GenBank mRNA | AK037507.1 | - | 13 - 14 | 1586 | 66 | |
| BC046414.1 | - | 7 - 8 | 648 | 66 | ||
| BC048939.1 | - | 11 - 12 | 1124 | 6439908 | 66 | |
More Information
Set Membership: |
|

Eogt
Genomic Map