Assay Details
| Gene Symbol: Papl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175319.4 | NP_780528.1 | 3 - 4 | 632 | 90 | |
| GenBank mRNA | AK049131.1 | - | 3 - 4 | 633 | 90 | |
| BC094908.1 | - | 3 - 4 | 528 | 30021730 | 90 | |
| BC132375.1 | - | 3 - 4 | 619 | 40130652 | 90 | |
| BC138307.1 | - | 3 - 4 | 618 | 8861327 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Papl
Genomic Map