Assay Details
Gene Symbol: E130012A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175332.3 | NP_780541.2 | 1 | 2063 | 83 | |
GenBank mRNA | AK087411.1 | - | 1 | 2050 | 83 | |
BC006054.1 | - | 1 | 1502 | 83 | ||
BC055770.1 | - | 1 | 2063 | 6403695 | 83 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051736 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051745 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051737 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051747 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051741 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051739 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051744 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051743 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051735 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051733 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051746 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051732 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051740 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161946 | AK087411.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161946 | BC055770.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161946 | NM_175332.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161946 | AK087411.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161946 | BC055770.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P161946 | NM_175332.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351085 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171005 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161946 | AK087411.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161946 | BC055770.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161946 | NM_175332.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161946 | AK087411.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161946 | BC055770.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P161946 | NM_175332.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051720 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051729 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051774 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051719 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28051770 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171005 | - | - |
Whole Mouse Genome G4122A | A_51_P161946 | AK087411.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161946 | BC055770.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161946 | NM_175332.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161946 | AK087411.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161946 | BC055770.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P161946 | NM_175332.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111517 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171005 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575693 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575691 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575694 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051736 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051740 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051739 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051747 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051741 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051732 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051735 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051737 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051744 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051746 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051745 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051743 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28051733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28051707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09644471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051728 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09607712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09298746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09433275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09128236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09644477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09062704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09666960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09183328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09555123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08988496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09575688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09298747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09575693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09575682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09183330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28051719 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390456 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390457 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390455 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426980_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390454 | - | - |
Mouse Genome 430 2.0 Array | 1426980_s_at | - | - |
Mouse Genome 430 2.0 Array | 1426980_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103551_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103551_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100033536.mm.1 | NM_175332.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100033536.mm.1 | NM_175332.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100033536.mm.1 | TC1100003648.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426980_PM_s_at | - | - |
Mouse Expression Set 430 | 1426980_s_at | - | - |
Murine Genome U74v2 Set | 160712_PM_r_at | - | - |
Murine Genome U74v2 Set | 160712_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1426980_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1426980_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426980_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426980_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 335619 | - | - |
ABI Mouse Genome Survey Microarray | 829427 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4210402 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004210402 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2629618 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2629618 | - | - |
mouse-6 v1.1 (Illumina) | scl39647.1.596_38-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4210402 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39647.1.596_38-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210402 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2963091 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2963091 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39647.1.596_38-S | - | - |
