Assay Details
Gene Symbol: Slc25a41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175333.3 | NP_780542.1 | 3 - 4 | 391 | 70 | |
GenBank mRNA | AK077111.1 | - | 3 - 4 | 392 | 70 | |
BC115588.1 | - | 4 - 5 | 490 | 40102382 | 70 | |
BC115589.1 | - | 3 - 4 | 384 | 40102384 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
