Assay Details
| Gene Symbol: Ano6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253813.1 | NP_001240742.1 | 8 - 9 | 1071 | 86 | |
| NM_175344.4 | NP_780553.2 | 7 - 8 | 1008 | 86 | ||
| GenBank mRNA | AK089300.1 | - | 7 - 8 | 921 | 87 | |
| AK159411.1 | - | 8 - 9 | 1022 | 86 | ||
| BC060732.1 | - | 7 - 8 | 1008 | 6409332 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089300.1 transcripts.
Set Membership: |
|

Ano6
Genomic Map