Assay Details
Gene Symbol: Zdhhc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175358.4 | NP_780567.1 | 10 - 11 | 1234 | 68 | |
GenBank mRNA | AK029137.1 | - | 10 - 11 | 974 | 68 | |
AK032922.1 | - | 10 - 11 | 955 | 68 | ||
AK077949.1 | - | 10 - 11 | 983 | 68 | ||
AK132304.1 | - | 10 - 11 | 1236 | 68 | ||
AK160360.1 | - | 10 - 11 | 974 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
