Assay Details
| Gene Symbol: Tmem139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175408.4 | NP_780617.2 | 3 | 805 | 81 | |
| GenBank mRNA | AK053212.1 | - | 1 | 2256 | 81 | |
| AK080737.1 | - | 1 | 81 | |||
| BC096053.1 | - | 1 | 488 | 3467410 | 81 | |
| CF585299.1 | - | 1 | 100 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995512 | CF585299.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995512 | AK053212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995512 | AK080737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995512 | BC096053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995512 | NM_175408.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431886 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112696 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995512 | CF585299.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995512 | AK053212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995512 | AK080737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995512 | BC096053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995512 | NM_175408.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24397150 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112696 | - | - |
| Whole Mouse Genome G4122A | A_51_P356667 | AK053212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356667 | AK080737.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123393 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139005 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05212901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05212893 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05459588 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05578302 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05578290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05212894 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05578344 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05212902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24397190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05840925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05432414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05208078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05358053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05432407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05212889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05212902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24397189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05212893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10537724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537726 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436556_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10537722 | - | - |
| Mouse Genome 430 2.0 Array | 1436556_at | - | - |
| Mouse Genome 430 2.0 Array | 1436556_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109218_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109218_at | - | - |
| Mouse Expression Set 430 | 1436556_PM_at | - | - |
| Mouse Expression Set 430 | 1436556_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436556_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436556_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 501695 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3780239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3780239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780239 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212753 | - | - |
| mouse-6 v1.1 (Illumina) | scl30125.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30125.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600822 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600822 | - | - |

Tmem139
Genomic Map