Assay Details
| Gene Symbol: Aldh4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175438.4 | NP_780647.3 | 10 - 11 | 1382 | 60 | |
| GenBank mRNA | AK044712.1 | - | 10 - 11 | 1178 | 60 | |
| BC024133.1 | - | 9 - 10 | 1100 | 60 | ||
| BC026589.1 | - | 5 - 6 | 594 | 60 | ||
| BC039281.1 | - | 10 - 11 | 1166 | 4975491 | 60 | |
| BC056226.1 | - | 10 - 11 | 1266 | 5064715 | 60 | |
| BC060650.1 | - | 9 - 10 | 2772 | 30354228 | 60 | |
| CK332989.1 | - | 4 - 5 | 492 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh4a1
Genomic Map