Assay Details
| Gene Symbol: Aldh4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175438.4 | NP_780647.3 | 2 - 3 | 399 | 79 | |
| GenBank mRNA | AK044712.1 | - | 2 - 3 | 195 | 79 | |
| BC024133.1 | - | 1 - 2 | 117 | 79 | ||
| BC039281.1 | - | 2 - 3 | 183 | 4975491 | 79 | |
| BC056226.1 | - | 2 - 3 | 283 | 5064715 | 79 | |
| BC060650.1 | - | 2 - 3 | 399 | 30354228 | 79 | |
| CA494657.1 | - | 2 - 3 | 436 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh4a1
Genomic Map