Assay Details
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175471.2 | NP_780680.1 | 4 - 5 | 724 | 65 | |
GenBank mRNA | AK220154.1 | - | 5 - 6 | 626 | 65 | |
BC043687.1 | - | 4 - 5 | 724 | 5136046 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138591 | AK053031.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138591 | AK220154.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004741 | BC043687.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004741 | AK053031.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004741 | AK220154.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004741 | BC004750.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004741 | NM_175471.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777352 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281040 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867973 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604036 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332269 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908261 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639204 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339640 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132839 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142885 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109125 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101731 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004741 | BC043687.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004741 | AK053031.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004741 | AK220154.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004741 | BC004750.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004741 | NM_175471.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138591 | AK053031.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138591 | AK220154.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188396 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188388 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188394 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188384 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188382 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188392 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188381 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188390 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188385 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101731 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142885 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109125 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05350351 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05350344 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05350381 | - | - |
Whole Mouse Genome G4122A | A_51_P143341 | AK053031.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P143341 | AK220154.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P18775 | BC043687.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P18775 | AK053031.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P18775 | AK220154.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P18775 | BC004750.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P18775 | NM_175471.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127848 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131866 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109125 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101731 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142885 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03902419 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03234749 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03902420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03888812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03546935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03679584 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04070512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03325003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03366169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03888813 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03573468 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03888814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03286138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04070514 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03234746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03210977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03386346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03234749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04052232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03234748 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04052233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03210973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03386347 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03366171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188384 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03679591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03210976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188429 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03366170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03546938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23188410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04052226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04070506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03325001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04052231 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10506629 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506630 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506632 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506635 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506633 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506637 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506631 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506636 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506634 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506628 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427423_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10506627 | - | - |
Mouse Genome 430 2.0 Array | 1427423_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427423_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400007720.mm.1 | NM_175471.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007720.mm.1 | TC0400001120.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007719.mm.1 | NM_175471.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007719.mm.1 | TC0400001120.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427423_at | - | - |
Mouse Expression Set 430 | 1427423_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427423_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427423_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427423_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427423_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 440438 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2350136 | - | - |
MouseWG-6 v2 Expression BeadChip | 2370484 | - | - |
MouseWG-6 v2 Expression BeadChip | 103130400 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2350136 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103130400 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2370484 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002370484 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002350136 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249086 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2666545 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2666545 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249086 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2466189 | - | - |
mouse-6 v1.1 (Illumina) | scl25167.9_473-S | - | - |
mouse-6 v1.1 (Illumina) | scl00230582.2_199-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002661.1_173-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2370484 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103130400 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2350136 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00230582.2_199-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002661.1_173-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370484 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350136 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103130400 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2964728 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244624 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2466190 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2964728 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244624 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2466190 | - | - |
