Assay Details
| Gene Symbol: Ppm1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175523.4 | NP_780732.1 | 5 - 6 | 1077 | 114 | |
| GenBank mRNA | AK044610.1 | - | 5 - 6 | 1147 | 114 | |
| BC092238.1 | - | 5 - 6 | 1077 | 30642218 | 114 | |
| BC138473.1 | - | 4 - 5 | 883 | 8861493 | 114 | |
| BC138474.1 | - | 4 - 5 | 883 | 8861494 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1k
Genomic Map