Assay Details
| Gene Symbol: Ddhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042719.2 | NP_001036184.1 | 12 - 13 | 2625 | 111 | |
| NM_001284399.1 | NP_001271328.1 | 13 - 14 | 2597 | 111 | ||
| NM_176845.5 | NP_789815.3 | 11 - 12 | 2523 | 111 | ||
| GenBank mRNA | AK014975.1 | - | 9 - 10 | 1309 | 111 | |
| AK048690.1 | - | 12 - 13 | 2083 | 111 | ||
| AK155294.1 | - | 13 - 14 | 2554 | 111 | ||
| AK170271.1 | - | 12 - 13 | 2237 | 111 | ||
| BC043475.1 | - | 10 - 11 | 1777 | 5365658 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddhd1
Genomic Map