| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400014422.mm.1 |
NM_001039106.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014422.mm.1 |
NM_001042719.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014422.mm.1 |
NM_001284399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014422.mm.1 |
NM_176845.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014422.mm.1 |
TC1400001906.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014423.mm.1 |
NM_001039106.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014423.mm.1 |
NM_001042719.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014423.mm.1 |
NM_001284399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014423.mm.1 |
NM_176845.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014423.mm.1 |
TC1400001906.mm.1 |
MTA TC |