Assay Details
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177323.3 | NP_796297.2 | 1 - 2 | 146 | 86 | |
GenBank mRNA | AK005284.1 | - | 1 - 2 | 179 | 86 | |
AK028707.1 | - | 1 - 2 | 177 | 86 | ||
AK133789.1 | - | 1 - 2 | 172 | 86 | ||
AK135334.1 | - | 1 - 2 | 167 | 86 | ||
AK136479.1 | - | 1 - 2 | 152 | 86 | ||
AK144144.1 | - | 1 - 2 | 168 | 86 | ||
AK160710.1 | - | 1 - 2 | 146 | 86 | ||
AK162664.1 | - | 1 - 2 | 123 | 86 | ||
BC065059.1 | - | 1 - 2 | 146 | 6401664 | 86 | |
BC145267.1 | - | 1 - 2 | 136 | 9053821 | 86 | |
BC150745.1 | - | 1 - 2 | 136 | 9087656 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK028707.1, AK135334.1, BC065059.1, BC145267.1, BC150745.1, NM_177323.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
