Assay Details
| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177324.2 | NP_796298.2 | 39 - 40 | 5550 | 76 | |
| GenBank mRNA | AK029035.1 | - | 6 - 7 | 893 | 76 | |
| AK029568.1 | - | 10 - 11 | 1553 | 76 | ||
| BC015069.1 | - | 15 - 16 | 2266 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sbf2
Genomic Map