Assay Details
| Gene Symbol: Vwa3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038906.1 | - | 4 - 5 | 694 | 65 | |
| AK050189.1 | - | 4 - 5 | 560 | 65 | ||
| AK076981.1 | - | 4 - 5 | 688 | 65 | ||
| BC049712.1 | - | 4 - 5 | 705 | 65 | ||
| BC147491.1 | - | 4 - 5 | 554 | 9087438 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vwa3b
Genomic Map