Assay Details
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290625.1 | NP_001277554.1 | 10 - 11 | 1489 | 73 | |
NM_001290626.1 | NP_001277555.1 | 9 - 10 | 1487 | 73 | ||
NM_177345.4 | NP_796319.1 | 10 - 11 | 1815 | 73 | ||
GenBank mRNA | AK027932.1 | - | 5 - 6 | 588 | 73 | |
AK042154.1 | - | 9 - 10 | 1477 | 73 | ||
AK052045.1 | - | 10 - 11 | 1483 | 73 | ||
AK132263.1 | - | 10 - 11 | 1479 | 73 | ||
BC027377.1 | - | 5 - 6 | 617 | 73 | ||
BC031579.1 | - | 5 - 6 | 617 | 73 | ||
BC043296.1 | - | 10 - 11 | 1714 | 73 | ||
BC090644.1 | - | 9 - 10 | 1389 | 30629263 | 73 | |
BC096618.1 | - | 10 - 11 | 1788 | 6392317 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
