Assay Details
| Gene Symbol: Mapkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177345.4 | NP_796319.1 | 1 - 2 | 411 | 84 | |
| GenBank mRNA | AK052045.1 | - | 1 - 2 | 79 | 84 | |
| AK148428.1 | - | 1 - 2 | 400 | 84 | ||
| BC043296.1 | - | 1 - 2 | 310 | 84 | ||
| BC096618.1 | - | 1 - 2 | 384 | 6392317 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052045.1, BC043296.1, BC096618.1, NM_177345.4 transcripts.
Set Membership: |
|

Mapkap1
Genomic Map