Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400016103.mm.1 |
NM_177366.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016103.mm.1 |
NM_177366.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016103.mm.1 |
TC0400001969.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016102.mm.1 |
NM_177366.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016102.mm.1 |
NM_177366.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016102.mm.1 |
TC0400001969.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016101.mm.1 |
NM_177366.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016101.mm.1 |
NM_177366.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016101.mm.1 |
TC0400001969.mm.1 |
MTA TC |